accessibility menu, dialog, popup

UserName

NH2SNOO

CAS RN 101-59-7

IUPAC Name: 4-[(4-nitrophenyl)sulfanyl]aniline
Synonyms: 4-amino-4'-nitrophenyl sulfide
Molecular Weight (g/mol): 246.28
Molecular Formula: C12H10N2O2S
InChi Key: ZBPKGHOGUVVDLF-UHFFFAOYSA-N
SMILES: NC1=CC=C(SC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)