accessibility menu, dialog, popup

UserName

CAS RN 101-10-0

IUPAC Name: 2-(3-chlorophenoxy)propanoic acid
Synonyms: cloprop
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: YNTJKQDWYXUTLZ-UHFFFAOYNA-N
SMILES: CC(OC1=CC=CC(Cl)=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results