accessibility menu, dialog, popup

UserName

H3CONBr

CAS RN 100367-39-3

IUPAC Name: 4-bromo-2-methoxypyridine
Synonyms: 4-bromo-2-methoxypyridine
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: YFTGMMXMLPTTAY-UHFFFAOYSA-N
SMILES: COC1=NC=CC(Br)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)