missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxypyridine, 98%

Catalog No. AAH6402606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6402606 5 g
AAH6402603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6402606 Supplier Thermo Scientific Chemicals Supplier No. H6402606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100367-39-3 | C6H6BrNO | 188.024 g/mol

4-Bromo-2-methoxypyridine used in the synhesis of Imidazo [1, 2-a] pyrimidines. Suzuki coupling between 4-methoxyphenylboronic acid and 4-bromo-2-methoxypyridine affords 1, 5-disubstituted pyridones. Starting material used in a synthesis of crown-ester-bipyridines and viologens via sodium or nickel reductive coupling, side chain oxidation and esterification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methoxypyridine used in the synhesis of Imidazo [1, 2-a] pyrimidines. Suzuki coupling between 4-methoxyphenylboronic acid and 4-bromo-2-methoxypyridine affords 1, 5-disubstituted pyridones. Starting material used in a synthesis of crown-ester-bipyridines and viologens via sodium or nickel reductive coupling, side chain oxidation and esterification.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

Chemical Identifiers

CAS 100367-39-3
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD08277268
InChI Key YFTGMMXMLPTTAY-UHFFFAOYSA-N
Synonym 2-methoxy-4-bromopyridine, 4-bromo-2-methoxy-pyridine, pyridine, 4-bromo-2-methoxy, pubchem6657, ksc494g1d, acmc-1c790, abbypharma ap-17-5965
PubChem CID 14062309
IUPAC Name 4-bromo-2-methoxypyridine
SMILES COC1=NC=CC(=C1)Br

Specifications

Melting Point 25°C to 26°C
Color White to Yellow
Boiling Point 87°C (15 mmHg)
Quantity 5 g
Beilstein 115199
Solubility Information Slightly soluble in water.
Formula Weight 188.02
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 4-Bromo-2-methoxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only