missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxypyridine, 97%

Catalog No. AC450000050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450000050 5 g
AC450000010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450000050 Supplier Thermo Scientific Chemicals Supplier No. 450000050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100367-39-3 | C6H6BrNO | 188.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100367-39-3
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
InChI Key YFTGMMXMLPTTAY-UHFFFAOYSA-N
Synonym 2-methoxy-4-bromopyridine, 4-bromo-2-methoxy-pyridine, pyridine, 4-bromo-2-methoxy, pubchem6657, ksc494g1d, acmc-1c790, abbypharma ap-17-5965
PubChem CID 14062309
IUPAC Name 4-bromo-2-methoxypyridine
SMILES COC1=NC=CC(=C1)Br

Specifications

Boiling Point 87.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 188.02
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methoxypyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only