missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Sulfate, Practical, Spectrum™ Chemical

Catalog No. 18609873
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609873 500 g Poly Bottle
18609871 2.5 kg Poly Bottle
18609872 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609873 Supplier Spectrum Chemical Mfg Cor Supplier No. C1326500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10026-24-1 | 281.10 g/mol

Chemical Identifiers

CAS 10026-24-1

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 500 g
Molecular Weight (g/mol) 281.10
Grade Practical
Packaging Poly Bottle
Chemical Name or Material Cobalt Sulfate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.