CAS RN 10025-82-8
IUPAC Name:
indium(3+) trichloride
Synonyms:
indium(III) chloride
Molecular Weight (g/mol):
221.17
Molecular Formula:
Cl3In
InChi Key:
PSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES:
[Cl-].[Cl-].[Cl-].[In+3]
1
–
5
of
5
results
1
Promotions Available
Indium(III) chloride, 99.995%, (trace metal basis)
Indium(III) chloride, 99.995%, (trace metal basis)
CAS: 10025-82-8 Molecular Formula: Cl3In Molecular Weight (g/mol): 221.18
2
Promotions Available
Indium(III) chloride, anhydrous, 99.99% (metals basis)
Indium(III) chloride, anhydrous, 99.99% (metals basis)
CAS: 10025-82-8 Molecular Formula: InCl3 MDL Number: MFCD00011058
3
Promotions Available
Indium(III) chloride, anhydrous, 98+%
Indium(III) chloride, anhydrous, 98+%
CAS: 10025-82-8 Molecular Formula: InCl3 MDL Number: MFCD00011058
4
Promotions Available
Indium(III) chloride, anhydrous, 99.999% (metals basis)
Indium(III) chloride, anhydrous, 99.999% (metals basis)
CAS: 10025-82-8 MDL Number: MFCD00011058
5
Indium(III) Chloride Anhydrous 99.0+%, TCI America™
CAS: 10025-82-8 Molecular Formula: Cl3In MDL Number: MFCD00011058