accessibility menu, dialog, popup

UserName

CAS RN 100124-06-9

IUPAC Name: {8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}boronic acid
Synonyms: 8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-ylboronic acid
Molecular Weight (g/mol): 212.01
Molecular Formula: C12H9BO3
InChi Key: ZXHUJRZYLRVVNP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC2=C1OC1=C2C=CC=C1
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)