missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran-4-boronic acid, 98+%

Catalog No. AAL1983006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1983006 5 g
AAL1983003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1983006 Supplier Thermo Scientific Chemicals Supplier No. L1983006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100124-06-9 | C12H9BO3 | 212.011 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100124-06-9
Molecular Formula C12H9BO3
Molecular Weight (g/mol) 212.011
MDL Number MFCD00092336
InChI Key ZXHUJRZYLRVVNP-UHFFFAOYSA-N
Synonym dibenzofuran-4-boronic acid, dibenzo b,d furan-4-ylboronic acid, 4-dibenzofuranyl boronic acid, 4-dibenzofuranboronic acid, 4-dibenzofuranboronicacid, 4-dibenzofuranylboronic acid, boronic acid, 4-dibenzofuranyl, dibenzo b,d furan-4-boronic acid, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid
PubChem CID 2734328
IUPAC Name dibenzofuran-4-ylboronic acid
SMILES B(C1=C2C(=CC=C1)C3=CC=CC=C3O2)(O)O

Specifications

Melting Point 286°C to 291°C
Quantity 5 g
Beilstein 1645426
Formula Weight 212.01
Percent Purity ≥98%
Chemical Name or Material Dibenzofuran-4-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only