missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran-4-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D45671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45671G Supplier TCI America Supplier No. D45671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100124-06-9
Molecular Formula C12H9BO3
MDL Number MFCD00092336
InChI Key ZXHUJRZYLRVVNP-UHFFFAOYSA-N
PubChem CID 2734328

Specifications

Melting Point 258°C
Color White
Quantity 1 g
Synonym dibenzofuran-4-boronic acid, dibenzo b,d furan-4-ylboronic acid, 4-dibenzofuranyl boronic acid, 4-dibenzofuranboronic acid, 4-dibenzofuranboronicacid, 4-dibenzofuranylboronic acid, boronic acid, 4-dibenzofuranyl, dibenzo b,d furan-4-boronic acid, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid
SMILES B(C1=C2C(=CC=C1)C3=CC=CC=C3O2)(O)O
IUPAC Name dibenzofuran-4-ylboronic acid
Molecular Weight (g/mol) 212.011
Formula Weight 212.01
Physical Form Crystalline Powder
Chemical Name or Material Dibenzofuran-4-boronic Acid (contains varying amounts of Anhydride)