accessibility menu, dialog, popup

UserName

H3CN

CAS RN 100-71-0

IUPAC Name: 2-ethylpyridine
Synonyms: 2-ethylpyridine
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: NRGGMCIBEHEAIL-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=N1
Percent Purity 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)