accessibility menu, dialog, popup

UserName

CAS RN 100-32-3

IUPAC Name: 1-nitro-4-[(4-nitrophenyl)disulfanyl]benzene
Synonyms: 1-nitro-4-[(4-nitrophenyl)disulfanyl]benzene
Molecular Weight (g/mol): 308.33
Molecular Formula: C12H8N2O4S2
InChi Key: KWGZRLZJBLEVFZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(SSC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results