missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100 μL
Clear, autoclavable tips; color-coded racks.
$228.93 - $2,708.09
Specifications
| Volume (Metric) | 100 μL |
|---|---|
| Color | Clear |
| Rack Color | Gray |
| Autoclavable | Autoclavable |
| Certifications/Compliance | Non-ANSI |
| Catalog Number | Mfr. No. | Sterility | Tip Style | Filtered | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sterility | Tip Style | Filtered | Price | Quantity | |||
|
14-222-921
|
Axygen™
ZTF100LRS |
Sterile | Maxymum Recovery, Round | Filtered |
N/A
|
|
|||
|
50-401-016
|
Axygen™
ZT100RS |
Sterile | Standard, Round | Non-filtered |
N/A
|
|
|||
|
14-222-907
|
Axygen™
ZT100R |
Non-sterile | Standard, Round | Non-filtered |
N/A
|
|
|||
|
50-401-026
|
Axygen™
ZTF100RS |
Sterile | Standard, Round | Filtered |
N/A
|
|
|||
Description
- Tip format: 96 tip and 384 tip
- Tip volume range: 25 μL to 200 μL
- Tip material: Clear polypropylene
- RNase-/DNase-/human gDNA-free
- Nonpyrogenic
- Available options:
- With or without aerosol-resistant filters
- Nonsterile or sterile
- Standard or Axygen™ Maxymum Recovery™ surface - Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
- Rack format: Non-ANSI (96) and ANSI/SLAS (384)
- Rack material: Polypropylene
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
| 100 μL | |
| Gray | |
| Non-ANSI | |
| Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head |
| Clear | |
| Autoclavable | |
| Rack | |
| Caliper Zymark |