Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100 μL
Catalog No. 14222921
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Maxymum Recovery, Round
Standard, Round
Filtered:
Filtered
Non-filtered
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Tip Style Filtered
14222921 Sterile Maxymum Recovery, Round Filtered
50401016 Sterile Standard, Round Non-filtered
14222907 Non-sterile Standard, Round Non-filtered
50401026 Sterile Standard, Round Filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14222921 Supplier Axygen™ Supplier No. ZTF100LRS
Clear, autoclavable tips; color-coded racks.
  • Tip format: 96 tip and 384 tip
  • Tip volume range: 25 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
  • Rack format: Non-ANSI (96) and ANSI/SLAS (384)
  • Rack material: Polypropylene
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Volume (Metric) 100 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Gray
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
Format Rack
Tip Style Maxymum Recovery, Round
Packaging Non-SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Product Line Caliper Zymark
Material Polyethylene
Show More Show Less