missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ZWITTERGENT™ 3-10 Detergent, Calbiochem™,

Supplier:  MilliporeSigma™ 6930215GM

 View more versions of this product

Catalog No. 6930215GM


Only null left

Description

Description

Specifications

Specifications

White
5 g
C15H33NO3S
n-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
307.49
≥99% (TLC)
15163-36-7
Glass Bottle
MFCD00036908
WKALLSVICJPZTM-UHFFFAOYSA-N
3-(decyldimethylazaniumyl)propane-1-sulfonate
161111

Chemical Identifiers

15163-36-7
307.49
WKALLSVICJPZTM-UHFFFAOYSA-N
161111
CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
C15H33NO3S
MFCD00036908
n-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
3-(decyldimethylazaniumyl)propane-1-sulfonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions