missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ZWITTERGENT™ 3-10 Detergent, Calbiochem™,

Catalog No. p-7082997
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6930215GM 5 g Glass Bottle
69302125GM 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6930215GM Supplier MilliporeSigma™ Supplier No. 6930215GM
Only null left

Chemical Identifiers

CAS 15163-36-7
Molecular Formula C15H33NO3S
Molecular Weight (g/mol) 307.49
MDL Number MFCD00036908
InChI Key WKALLSVICJPZTM-UHFFFAOYSA-N
Synonym n-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
PubChem CID 161111
IUPAC Name 3-(decyldimethylazaniumyl)propane-1-sulfonate
SMILES CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥99% (TLC)