missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ ZWITTERGENT™ 3-10 Detergent, Calbiochem™,
Supplier: MilliporeSigma™ 69302125GM
| Quantity | 25 g |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| 15163-36-7 | |
| 307.49 | |
| WKALLSVICJPZTM-UHFFFAOYSA-N | |
| 161111 | |
| CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O |
| C15H33NO3S | |
| MFCD00036908 | |
| n-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate | |
| 3-(decyldimethylazaniumyl)propane-1-sulfonate |
Specifications
| White | |
| 25 g | |
| C15H33NO3S | |
| n-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate | |
| CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O | |
| 307.49 | |
| ≥99% (TLC) |
| 15163-36-7 | |
| Glass Bottle | |
| MFCD00036908 | |
| WKALLSVICJPZTM-UHFFFAOYSA-N | |
| 3-(decyldimethylazaniumyl)propane-1-sulfonate | |
| 161111 |