missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZSCAN9 Polyclonal Antibody

Description
Immunogen sequence: ELVLRKDCPK IVEPHGKMFN EQTWEVSQQD PSHGEVGEHK DRIERQWGNL LGEGQHKCDE Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 30%.
ZNF193 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 5 C2H2-type zinc fingers and 1 SCAN box domain. ZNF193 may be involved in transcriptional regulation.
Specifications
Specifications
| Antigen | ZSCAN9 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZSCAN9 |
| Gene Accession No. | O15535 |
| Gene Alias | Cell proliferation-inducing gene 12 protein; cell proliferation-inducing protein 12; PIG12; PRD51; zinc finger and SCAN domain containing 9; zinc finger and SCAN domain-containing protein 9; Zinc finger protein 193; ZNF193; ZSCAN9 |
| Gene Symbols | ZSCAN9 |
| Show More |