missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZPR1 Monoclonal Antibody (LG1)
GREENER_CHOICE

Catalog No. PIMA113003
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA113003 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA113003 Supplier Invitrogen™ Supplier No. MA113003
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

MA1-13003 detects ZPR1 in human and mouse samples. MA1-13003 has been successfully been used in Immunofluorescence and Western Blot applications, where a band of ~50 kDa is detected. The immunogen used to produce MA1-13003 was mouse ZPR1 protein.

The protein encoded by this gene is found in the cytoplasm of quiescent cells but translocates to the nucleolus in proliferating cells. The encoded protein interacts with survival motor neuron protein (SMN1) to enhance pre-mRNA splicing and to induce neuronal differentiation and axonal growth. Defects in this gene or the SMN1 gene can cause spinal muscular atrophy. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Nov 2015].

Specifications

Antigen ZPR1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone LG1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene ZPR1
Gene Accession No. O75312, Q62384
Gene Alias AI303781; fc17g10; hypothetical protein LOC504523; RGD1562173; si:rp71-56i13.4; wu:fc17g10; Zfp259; zgc:56478; Zinc finger protein 259; zinc finger protein ZPR1; ZNF259; Zpr1; ZPR1 zinc finger
Gene Symbols ZPR1
Host Species Mouse
Immunogen murine ZPR1 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22687, 8882
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less