missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZPR1 Monoclonal Antibody (K1E008_6E12), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555625
Description
Sequence of this protein is as follows: VRYTLSVRAL EDMNREVVKT DSAATRIPEL DFEIPAFSQK GALTTVEGLI TRAISGLEQD QPARRANKDA TAERIDEFIV KLKELKQVAS PFTLIIDDPS GNSFVENPHA PQKDDALVIT HYNRTRQQEE MLGLQEEAPA EKPEEEDLRN EVLQFSTNCP ECNAPAQTNM KLVQIPHFKE VIIMATNCEN CGHRTNEVKS GGAVEPLGTR ITLHITDASD MTRDLLKSET CSVEIPELEF ELGMAVLGGK FTTLEGLLKD IRELVTKNPF TLGDSSNPGQ TERLQEFSQK MDQIIEGNMK AHFIMDDPAG NSYLQNVYAP EDDPEMKVER
The protein encoded by this gene is found in the cytoplasm of quiescent cells but translocates to the nucleolus in proliferating cells. The encoded protein interacts with survival motor neuron protein (SMN1) to enhance pre-mRNA splicing and to induce neuronal differentiation and axonal growth. Defects in this gene or the SMN1 gene can cause spinal muscular atrophy. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Nov 2015].Specifications
| ZPR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O75312, Q62384 | |
| ZPR1 | |
| Recombinant human ZPR1 (immunogen range is 101-430). | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K1E008_6E12 | |
| Unconjugated | |
| ZPR1 | |
| AI303781; fc17g10; hypothetical protein LOC504523; RGD1562173; si:rp71-56i13.4; wu:fc17g10; Zfp259; zgc:56478; Zinc finger protein 259; zinc finger protein ZPR1; ZNF259; Zpr1; ZPR1 zinc finger | |
| Mouse | |
| Protein A | |
| RUO | |
| 22687, 500989, 8882 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |