missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZPR1 Monoclonal Antibody (K1E008_6E12), Invitrogen™

Catalog No. MA556005
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556005 200 μL
MA555625 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556005 Supplier Invitrogen™ Supplier No. MA556005
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: VRYTLSVRAL EDMNREVVKT DSAATRIPEL DFEIPAFSQK GALTTVEGLI TRAISGLEQD QPARRANKDA TAERIDEFIV KLKELKQVAS PFTLIIDDPS GNSFVENPHA PQKDDALVIT HYNRTRQQEE MLGLQEEAPA EKPEEEDLRN EVLQFSTNCP ECNAPAQTNM KLVQIPHFKE VIIMATNCEN CGHRTNEVKS GGAVEPLGTR ITLHITDASD MTRDLLKSET CSVEIPELEF ELGMAVLGGK FTTLEGLLKD IRELVTKNPF TLGDSSNPGQ TERLQEFSQK MDQIIEGNMK AHFIMDDPAG NSYLQNVYAP EDDPEMKVER

The protein encoded by this gene is found in the cytoplasm of quiescent cells but translocates to the nucleolus in proliferating cells. The encoded protein interacts with survival motor neuron protein (SMN1) to enhance pre-mRNA splicing and to induce neuronal differentiation and axonal growth. Defects in this gene or the SMN1 gene can cause spinal muscular atrophy. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Nov 2015].

Specifications

Antigen ZPR1
Applications Western Blot
Classification Monoclonal
Clone K1E008_6E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene ZPR1
Gene Accession No. O75312, Q62384
Gene Alias AI303781; fc17g10; hypothetical protein LOC504523; RGD1562173; si:rp71-56i13.4; wu:fc17g10; Zfp259; zgc:56478; Zinc finger protein 259; zinc finger protein ZPR1; ZNF259; Zpr1; ZPR1 zinc finger
Gene Symbols ZPR1
Host Species Mouse
Immunogen Recombinant human ZPR1 (immunogen range is 101-430).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22687, 500989, 8882
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less