Learn More
Motic Swift Line Zoom Stereo Microscope
Catalog No. S01437
Encompass
Change view
Click to view available options
Includes:
Zoom Stereo Microscope
Zoom Stereo Microscope w/ 8 in. Tablet
Zoom Stereo Microscope with HD Camera
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S01437 Zoom Stereo Microscope
S01438 Zoom Stereo Microscope w/ 8 in. Tablet
S01439 Zoom Stereo Microscope with HD Camera
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S01437 Supplier Motic Swift Line Supplier No. M30TZSM99CL
Only null left
The M30TZ is ideal for laboratory, industrial and educational use. This stereo microscope features zoom objectives with a continuous magnification range of 7.5X to 40X, offering added versatility that is not an option with fixed power stereo microscopes.
  • Paired widefield eyepieces are 45° inclined, with diopter adjustment. Interpupillary adjustment from 55 to 75 mm.
  • Trinocular Port accepts video adapter.
  • Interlocked eyepiece tubes with diopter for increased focus acuity.
  • Viewing head rotates 360°. Locks in place by tightening thumb screw.
  • Paired objectives are parfocalled, parcentered, and achromatic.
  • Magnification changes by rotating magnification dial.
  • 3W LED illumination provides bright, white light with virtually no heat.
  • Top light adjusts for optimal lighting angles.
  • Variable transbase illumination allows user to control brightness.
  • Easy-to-replace LED illumination is energy-efficient and rarely requires bulb replacement.
  • Dual illumination switches allow for 3 light settings.
  • Rack and pinion focusing with slip clutch and tension adjustment.
  • Slip-clutch system prevents users from damaging the focusing mechanism.
  • 100 mm reversible black/white plastic contrast plate and 100 mm frosted glass stage plate.
  • Locked-on stage clips.
  • Rugged, cast metal frame.
  • Heavy-duty post mount with locking support collar, permits up or down adjustment of viewing head to accommodate different sized objects on stage.
  • Microscopes with 8 in. tablet or HD Camera available.
Specifications
Includes Zoom Stereo Microscope
Magnification Power 7.5X to 40X
Objective Types Achromatic
Head Style Trinocular
Lighting Type LED
Eyepiece WF 10X