Learn More
Zonisamide, Thermo Scientific™
$154.16 - $484.48
Chemical Identifiers
| CAS | 68291-97-4 |
|---|---|
| Molecular Formula | C8H8N2O3S |
| Molecular Weight (g/mol) | 212.22 |
| MDL Number | MFCD00865316 |
| InChI Key | UBQNRHZMVUUOMG-UHFFFAOYSA-N |
| IUPAC Name | (1,2-benzoxazol-3-yl)methanesulfonamide |
| SMILES | NS(=O)(=O)CC1=NOC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC465040010
|
N/A | 1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC465042500
|
N/A | 250 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 68291-97-4 | |
| 212.22 | |
| UBQNRHZMVUUOMG-UHFFFAOYSA-N | |
| NS(=O)(=O)CC1=NOC2=CC=CC=C12 |
| C8H8N2O3S | |
| MFCD00865316 | |
| (1,2-benzoxazol-3-yl)methanesulfonamide |
Specifications
| 68291-97-4 | |
| MFCD00865316 | |
| UBQNRHZMVUUOMG-UHFFFAOYSA-N | |
| (1,2-benzoxazol-3-yl)methanesulfonamide | |
| 212.22 |
| C8H8N2O3S | |
| Solubility aq. soln.: .. Other solubilities: sparingly soluble in chloroform, n-hexane, soluble in methanol, ethanol, ethyl acetate and, acetic acid | |
| NS(=O)(=O)CC1=NOC2=CC=CC=C12 | |
| 212.22 | |
| Zonisamide |
Safety and Handling
GHS H Statement Harmful if swallowed.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
RUO – Research Use Only