missing translation for 'onlineSavingsMsg'
Learn More

Zonisamide, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 465040010

Quantity 1 g

 View more versions of this product

Catalog No. AC465040010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

68291-97-4
212.22
UBQNRHZMVUUOMG-UHFFFAOYSA-N
NS(=O)(=O)CC1=NOC2=CC=CC=C12
C8H8N2O3S
MFCD00865316
(1,2-benzoxazol-3-yl)methanesulfonamide

Specifications

Zonisamide
C8H8N2O3S
MFCD00865316
UBQNRHZMVUUOMG-UHFFFAOYSA-N
(1,2-benzoxazol-3-yl)methanesulfonamide
212.22
68291-97-4
1 g
Solubility aq. soln.: .. Other solubilities: sparingly soluble in chloroform, n-hexane, soluble in methanol, ethanol, ethyl acetate and, acetic acid
NS(=O)(=O)CC1=NOC2=CC=CC=C12
212.22
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

May cause drowsiness or dizziness.

Suspected of damaging fertility.

Suspected of damaging the unborn child.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions