missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Zonisamide | |
| 165°C | |
| C8H8N2O3S | |
| 1 g | |
| UBQNRHZMVUUOMG-UHFFFAOYSA-N | |
| (1,2-benzoxazol-3-yl)methanesulfonamide | |
| 5734 | |
| 212.22 | |
| Crystalline Powder |
| 68291-97-4 | |
| White-Yellow | |
| MFCD00865316 | |
| zonisamide, zonegran, exceglan, excegram, 1,2-benzisoxazole-3-methanesulfonamide, zonisamida, zonisamidum, zonisamidum latin, zonisamida spanish, excegran | |
| NS(=O)(=O)CC1=NOC2=CC=CC=C12 | |
| 212.22 | |
| CHEBI:10127 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 68291-97-4 | |
| 212.22 | |
| UBQNRHZMVUUOMG-UHFFFAOYSA-N | |
| 5734 | |
| (1,2-benzoxazol-3-yl)methanesulfonamide |
| C8H8N2O3S | |
| MFCD00865316 | |
| zonisamide, zonegran, exceglan, excegram, 1,2-benzisoxazole-3-methanesulfonamide, zonisamida, zonisamidum, zonisamidum latin, zonisamida spanish, excegran | |
| CHEBI:10127 | |
| NS(=O)(=O)CC1=NOC2=CC=CC=C12 |
Safety and Handling
RTECSNumber : DE4930000
TSCA : No