missing translation for 'onlineSavingsMsg'
Learn More

Zonisamide 98.0+%, TCI America™

Catalog No. p-7137998
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0026200MG 200 mg
Z00261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z0026200MG Supplier TCI America Supplier No. Z0026200MG

Chemical Identifiers

CAS 68291-97-4
Molecular Formula C8H8N2O3S
Molecular Weight (g/mol) 212.22
MDL Number MFCD00865316
InChI Key UBQNRHZMVUUOMG-UHFFFAOYSA-N
Synonym zonisamide, zonegran, exceglan, excegram, 1,2-benzisoxazole-3-methanesulfonamide, zonisamida, zonisamidum, zonisamidum latin, zonisamida spanish, excegran
PubChem CID 5734
ChEBI CHEBI:10127
IUPAC Name (1,2-benzoxazol-3-yl)methanesulfonamide
SMILES NS(=O)(=O)CC1=NOC2=CC=CC=C12

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 200 mg
Formula Weight 212.22
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Zonisamide
RTECSNumber DE4930000
TSCA No