missing translation for 'onlineSavingsMsg'
Learn More

ZoBell's Solution, Oxidation-Reduction Potential (ORP) Standard, Ricca Chemical

Catalog No. p-3252652
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
988016 500 mL Amber Poly Bottle
988032 1 L Amber Poly Bottle
98801 4 L Black Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 988016 Supplier Ricca Chemical Company Supplier No. 988016
Only null left

Specifications

Chemical Name or Material ZoBell's Solution
Packaging Amber Poly Bottle
Name Note Oxidation-Reduction Potential (ORP) Standard
CAS Min % 97.0, 0.73, 0.23, 0.11
CAS Max % 100.0, 0.76, 0.24, 0.11
CAS 13746-66-2, 7447-40-7, 14459-95-1, 7732-18-5
Physical Form Liquid
Boiling Point 100°C
Quantity 500 mL