missing translation for 'onlineSavingsMsg'
Learn More

ZoBell's Solution, Modified, Oxidation-Reduction Potential (ORP) Standard, Ricca Chemical

Catalog No. p-3252651
Change view
Click to view available options
Quantity:
120 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98814 120 mL
988132 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 98814 Supplier Ricca Chemical Company Supplier No. 98814
Only null left

Specifications

Chemical Name or Material ZoBell's Solution
Packaging Amber Poly Bottle
Name Note Oxidation-Reduction Potential (ORP) Standard
CAS Min % 97.0, 0.73, 0.72, 7.0E-4
CAS Max % 100.0, 0.76, 0.75, 7.0E-4
CAS 13746-66-2, 14459-95-1, 7447-40-7, 7732-18-5
Physical Form Liquid
Color Yellow
Boiling Point 100°C
Quantity 120 mL