missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF645 Monoclonal Antibody (K92039_7G8), Invitrogen™

Catalog No. MA555671
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555671 50 μL
MA556051 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555671 Supplier Invitrogen™ Supplier No. MA555671
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: FTRKHGNLTV DHIQNNSDSG AKKPTPPDYY PECQSQPAVS SPHHIIPQKQ HYAPPPSPSS PVNHQMPYPP QDVVTPNSVR SQVPALTTTY DPSSGYIIVK VPPDMNSPPL RAPQSQNGNP

ZNF645 contains 1 RING-type zinc finger, which is probably involved in mediating protein-protein interactions.

Specifications

Antigen ZNF645
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K92039_7G8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene ZNF645
Gene Accession No. Q8N7E2
Gene Alias Cbl proto-oncogene-like protein 2; CBLL2; c-Cbl-like protein 2; CT138; E3 ubiquitin ligase; E3 ubiquitin-protein ligase CBLL2; E3 ubiquitin-protein ligase ZNF645; HAKAIL; RING-type E3 ubiquitin transferase ZNF645; Zinc finger protein 645; ZNF645
Gene Symbols ZNF645
Host Species Mouse
Immunogen Recombinant human CBLL2 (immunogen range is 231-350).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158506
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less