missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF597 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532066
Description
Recommended positive controls: A431. Predicted reactivity: Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in transcriptional regulation.Chemical Identifiers
| ZNF597 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| ZNF597 | |
| HIT-4; zinc finger protein 597; ZNF597 | |
| ZNF597 | |
| Primary | |
| Antigen affinity chromatography |
| 0.58 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96LX8 | |
| 146434 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF597. The exact sequence is proprietary. | |
| Antibody |
Specifications
| ZNF597 | |
| Polyclonal | |
| Unconjugated | |
| ZNF597 | |
| HIT-4; zinc finger protein 597; ZNF597 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 146434 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.58 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96LX8 | |
| ZNF597 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF597. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |