missing translation for 'onlineSavingsMsg'
Learn More

Saccharin, Powder, FCC, 98-101%, Spectrum™ Chemical

Catalog No. 18606127
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606127 2.5 kg Poly Bottle
18606125 125 g Poly Bottle
18606126 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606127 Supplier Spectrum Chemical Mfg Cor Supplier No. S10032.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1003, 81-07-2, C7H5NO3S

Spectrum™ Chemical Saccharin, Powder, FCC is used as an artificial sweetener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 81-07-2
Molecular Formula C7H5NO3S
Molecular Weight (g/mol) 183.18
InChI Key CVHZOJJKTDOEJC-UHFFFAOYSA-N
IUPAC Name 2,3-dihydro-1λ6,2-benzothiazole-1,1,3-trione
SMILES O=C1NS(=O)(=O)C2=CC=CC=C12

Specifications

CAS Max % 1
Ignition Residue 0.002
Loss on Drying 0.01
Melting Range 226-230°C
Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 98 to 101%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Saccharin, Powder