accessibility menu, dialog, popup

UserName

CAS RN 81-07-2

IUPAC Name: 2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
Synonyms: saccharine saccharimide benzosulfimide o-sulfobenzoic acid imide benzoic sulphimide o-benzosulfimide benzoic acid sulfimide benzoic sulfimide benzosulphimide o-sulfobenzimide
Molecular Weight (g/mol): 183.18
Molecular Formula: C7H5NO3S
InChi Key: CVHZOJJKTDOEJC-UHFFFAOYSA-N
SMILES: O=C1NS(=O)(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (10)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
Grade 
  • (3)
  • (4)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (7)
  • (2)
  • (6)
  • (10)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (3)
  • (4)
  • (2)
  • (3)