missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF555 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112803
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112803 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112803 Supplier Invitrogen™ Supplier No. PA5112803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

ZNF555 Polyclonal Antibody for Western Blot, IHC (P), ELISA

ZNF555 contains 15 C2H2-type zinc fingers and 1 KRAB domain. It belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.

Specifications

Antigen ZNF555
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZNF555
Gene Accession No. Q8NEP9
Gene Alias MGC26707; Zinc finger protein 555; ZNF555
Gene Symbols ZNF555
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein 555 protein (1-200AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148254
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less