missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF50 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552299
Description
Immunogen sequence: TQVLDKRGWD PGAEPTEASC KQSDLGESEP SNVTETLMGG VSLGPAFVKA CEPEGSSERS GLSGEIWTKS VTQQIHFKKT SGPYKDVPTD QRGRESGASR NSSSAWPNLT SQ Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 46%.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZSCAN22 (zinc finger and SCAN domain-containing protein 22), also known as HKR2 or ZNF50 (zinc finger protein 50), is a 491 amino acid nuclear protein. Belonging to the Krueppel C2 H2-type zinc-finger protein family, ZSCAN22 contains eight C2 H2-type zinc fingers and one SCAN box domain. ZSCAN22 may be involved in transcriptional regulation, and becomes phosphorylated upon DNA damage.Specifications
| ZNF50 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN22 | |
| AW227621; D530006B18Rik; GLI-Kruppel family member HKR2; HKR2; Krueppel-related zinc finger protein 2; LOC102553002; Protein HKR2; zinc finger and SCAN domain containing 22; zinc finger and SCAN domain-containing protein 22; Zinc finger protein 50; zinc finger protein 8-like; ZNF50; ZSCAN22 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 342945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P10073 | |
| ZSCAN22 | |
| Recombinant protein corresponding to Human ZNF50. Recombinant protein control fragment (Product #RP-89416). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |