missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF445 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552322
Description
Immunogen sequence: QPKGNPVAAP TGDDLQSKTN KFILNQEPLE EAETLAVSSG CPATSVSEGI GLRESFQQKS RQKDQCENPI QVRVKKEETN FSHRTGKDSE VSGSNSLDLK HVTYLRVSGR KESLKHGCGK HFRMSSHHYD YKKYGK Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 47%.
ZNF445 gene ontology annotations related to this gene include DNA-binding transcription factor activity; metal ion binding; nucleic acid binding.Specifications
| ZNF445 | |
| Polyclonal | |
| Unconjugated | |
| ZNF445 | |
| AW610627; mszf29; mszf50; RGD1559144; Zfp445; Zinc finger protein 168; zinc finger protein 445; Zinc finger protein with KRAB and SCAN domains 15; ZKSCAN15; ZNF168; Znf445; ZSCAN47 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 353274 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P59923 | |
| ZNF445 | |
| Recombinant protein corresponding to Human ZNF445. Recombinant protein control fragment (Product #RP-89419). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |