missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF419 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531951 Supplier Invitrogen™ Supplier No. PA531951
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May be involved in transcriptional regulation.

Specifications

Antigen ZNF419
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZNF419
Gene Accession No. Q96HQ0
Gene Alias ZAPHIR; zinc finger protein 419; zinc finger protein 419A; ZNF419; ZNF419A
Gene Symbols ZNF419
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human ZNF419. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79744
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less