missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553673
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553673 Supplier Invitrogen™ Supplier No. PA553673
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: RSYRKAKSSH PPGTCPFYEE LDSLMRARAA VRAMGTVREA AGLPRCGQSS AETDAQEAWG EVANEDAVKP STLCPKAPDM GFEMRHEDED QISEQDIFEG LPGALSKCPT EAVCQPLDWG EDSENENEDE GQWG Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 61%.

May be involved in transcriptional regulation.

Specifications

Antigen ZNF31
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZSCAN20
Gene Accession No. P17040
Gene Alias C130001F22; KOX29; Zfp31; ZFP-31; zinc finger and SCAN domain containing 20; zinc finger and SCAN domain-containing protein 20; zinc finger and SCAN domains 20; zinc finger protein 31; Zinc finger protein 360; zinc finger protein KOX29; zinc finger with KRAB and SCAN domains 20; Zkscan20; ZNF31; ZNF360; ZSCAN20
Gene Symbols ZSCAN20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF31. Recombinant protein control fragment (Product #RP-91317).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7579
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less