missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa536353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536353 Supplier Invitrogen™ Supplier No. PA536353

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 73 kDa. Purity is >95% by SDS-PAGE.

ZNF23 May be involved in transcriptional regulation. May have a role in embryonic development. Belongs to the krueppel C2H2-type zinc-finger protein family.

Specifications

Antigen ZNF23
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF23
Gene Accession No. P17027
Gene Alias B230354B21Rik; KOX16; kruppel-like zinc finger factor X31; Zfp23; Zfp612; zinc finger protein 23; zinc finger protein 23 (KOX 16); zinc finger protein 32; zinc finger protein 359; Zinc finger protein 612; Zinc finger protein KOX16; ZNF23; ZNF359; ZNF612
Gene Symbols Zfp612, ZNF23
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 150-200 of human ZNF23.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234725, 307839, 7571
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less