missing translation for 'onlineSavingsMsg'
Learn More

ZNF198 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501561035
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501561035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501561035 Supplier Bethyl Laboratories Inc Supplier No. A301710A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan

The protein encoded by this gene is a zinc finger protein that may act as a transcription factor. The encoded protein may be part of a BHC histone deacetylase complex. Translocation of this gene with the fibroblast growth factor receptor-1 gene (FGFR1) results in a fusion gene, which may be a cause of stem cell leukemia lymphoma syndrome (SCLL). Several transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen ZNF198
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZMYM2
Gene Accession No. Q9UBW7
Gene Alias ZNF198
Gene Symbols ZMYM2
Host Species Rabbit
Immunogen Between 975 and 1025
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7750
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less