missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconyl-SPADNS Reagent, for Fluoride Analysis, Ricca Chemical
For Analysis of Fluoride
$187.56 - $310.25
Specifications
| Chemical Name or Material | Zirconyl-SPADNS Reagent |
|---|---|
| Color | Red |
| CAS | 7732-18-5,7647-01-0,23647-14-5,13520-92-8 |
| CAS Min % | 69.0,28.62,0.07,0.01 |
| CAS Max % | 72.0,29.79,0.07,0.01 |
Specifications
| Zirconyl-SPADNS Reagent | |
| 7732-18-5,7647-01-0,23647-14-5,13520-92-8 | |
| 72.0,29.79,0.07,0.01 | |
| Laboratory | |
| <1 |
| Red | |
| 69.0,28.62,0.07,0.01 | |
| dihydrogen oxide, dihydrogen monoxide | |
| Amber Glass Bottle | |
| Liquid |