missing translation for 'onlineSavingsMsg'
Learn More

Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf

Catalog No. AC222631000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222631000 100 g Glass bottle
AC222635000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC222631000 Supplier Thermo Scientific Chemicals Supplier No. 222631000

CAS: 14985-18-3 | N2O7Zr | 231.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14985-18-3
Molecular Formula N2O7Zr
Molecular Weight (g/mol) 231.23
MDL Number MFCD00149896
InChI Key JWBWFTANBHYHBC-UHFFFAOYSA-N
Synonym zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis
PubChem CID 16211674
IUPAC Name oxozirconium(2+);dinitrate;hydrate
SMILES [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O

Specifications

Color White
Physical Form Chunks and/or Powder
Quantity 100 g
Assay Percent Range 99.50%
Linear Formula ZrO(NO3)2·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 231.23
Percent Purity 99.5%
Packaging Glass bottle
Chemical Name or Material Zirconyl(IV) nitrate hydrate, 99.5%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only