missing translation for 'onlineSavingsMsg'
Learn More

Zirconocene Chloride Hydride 96.0+%, TCI America™

Catalog No. p-7137991
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z00101G 1 g
Z00105G 5 g
Z001025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Z00101G Supplier TCI America Supplier No. Z00101G
Only null left

Chemical Identifiers

CAS 37342-97-5
Molecular Formula C10H11ClZr-2
Molecular Weight (g/mol) 257.872
MDL Number MFCD02089401
InChI Key VPQMECJLNPUNGN-UHFFFAOYSA-M
Synonym zirconocene chloride hydride
PubChem CID 131718033
IUPAC Name cyclopenta-1,3-diene;hydride;zirconium(2+);chloride
SMILES [H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2]

Specifications

Quantity 1 g
UN Number 3131
Formula Weight 257.87
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zirconocene Chloride Hydride