Learn More
Zirconium, plasma standard solution, Specpure™ Zr 10,000μg/mL
$259.40 - $259.40
Description
Zirconium plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| MFCD00011303 |
Specifications
| Liquid | |
| MFCD00011303 | |
| Miscible with water | |
| Specpure |
| ZrCl2O in 5% HCl | |
| UN1789 | |
| 100 mL | |
| Zirconium plasma standard solution, Zr 10000μg/mL |
Safety and Handling
GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID
EINECSNumber : 231-595-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only