missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) tert-butoxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. AA3954609
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3954609 10 g
AA3954604 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3954609 Supplier Thermo Scientific Chemicals Supplier No. 03954609

It finds it application as a reagent for the preparation of heterometallic tin(II) or lead(II) zirconium alkoxides. It is used as a catalyst in cyanation process. Zirconium tert-butoxide (ZTB) precursor is used to deposit thin films of Zirconia and other zirconium containing films by atomic layer deposition and chemical vapor deposition methods. Zirconium oxide thin films can also be grown at low temperatures, ranging from 150°C to 300°C, by the presence of moisture along with ZTB or by UV-enhanced atomic layer deposition (UV-ALD) process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2081-12-1
Molecular Formula C16H40O4Zr
Molecular Weight (g/mol) 387.72
MDL Number MFCD00075085
InChI Key WRMYASGYMABHCC-UHFFFAOYSA-N
Synonym zirconium iv tert-butoxide, 2-methylpropan-2-ol; zirconium
PubChem CID 25203936
IUPAC Name 2-methylpropan-2-ol;zirconium
SMILES CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.[Zr]

Specifications

Color Colorless
Density 0.985 g/mL
Boiling Point 90°C (5 mmHg)
Flash Point 85°C (185°F)
Linear Formula Zr[OC(CH3)3]4
Quantity 10 g
Beilstein 3681870
Sensitivity Light and moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 383.68
Percent Purity 99.99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Zirconium(IV) tert-butoxide
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only