missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™

Catalog No. AC427931000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427931000 100 mL AcroSeal™ Glass Bottle
AC427938000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427931000 Supplier Thermo Scientific Chemicals Supplier No. 427931000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23519-77-9 | C12H28O4Zr | 327.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23519-77-9 , 71-23-8
Molecular Formula C12H28O4Zr
Molecular Weight (g/mol) 327.58
MDL Number MFCD00015307
InChI Key XPGAWFIWCWKDDL-UHFFFAOYSA-N
Synonym zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
PubChem CID 90139
SMILES CCCO[Zr](OCCC)(OCCC)OCCC

Specifications

Color Amber to Colorless
Density 1.0440g/mL
Flash Point 23°C
Assay Percent Range 27.0 to 29% (ZrO2)
Packaging AcroSeal™ Glass Bottle
Linear Formula Zr(OCH2CH2CH3)4
Quantity 100 mL
Specific Gravity 1.044
IUPAC Name zirconium(4+) tetrakis(propan-1-olate)
Formula Weight 327.57
Percent Purity ca. 70%
Physical Form Slightly Viscous Liquid
Chemical Name or Material Zirconium(IV) propoxide, ca. 70%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 245-711-9

RUO – Research Use Only