accessibility menu, dialog, popup

UserName

CAS RN 23519-77-9

IUPAC Name: tetrapropoxyzirconium
Synonyms: tetrapropoxyzirconium
Molecular Weight (g/mol): 327.58
Molecular Formula: C12H28O4Zr
InChi Key: XPGAWFIWCWKDDL-UHFFFAOYSA-N
SMILES: CCCO[Zr](OCCC)(OCCC)OCCC
Physical Form 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (7)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)