missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) Oxide Chloride Octahydrate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1089170100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1089170100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1089170100 Supplier Technidata Supplier No. 1.08917.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity EMSURE™ grade reagent suitable for analytical laboratory applications.

  • Formula: ZrOCl2 ¹ 8H2O
  • Assay (gravimetric): ≥97.0%
  • Physical State: Solid
  • Density: 1.91 g/cm³
  • Properties: Hygroscopic
  • Hazard Classification: Skin corrosion (Category 1B); causes severe skin burns and eye damage
  • Storage: 15°C to 25°C

Chemical Identifiers

CAS 13520-92-8
Molecular Formula Cl2H16O9Zr
Molecular Weight (g/mol) 322.24
MDL Number MFCD00149898
InChI Key SDFOCNSOSARAKC-UHFFFAOYSA-L
Synonym zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
PubChem CID 159678
IUPAC Name oxozirconium;octahydrate;dihydrochloride
SMILES O.Cl[Zr](Cl)=O

Specifications

pH 1
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥97.0%
Linear Formula ZrOCl2 · 8H2O
UN Number UN 3260
InChI Formula InChI=1S/2ClH.8H2O.O.Zr/h2*1H;8*1H2;;
Formula Weight 322.28
Concentration or Composition (by Analyte or Components) Assay (gravimetrically): ≥ 97.0%, Sulfate (SO4): ≤ 0.015%, Heavy metals (as Pb): ≤ 0.001%, Fe (Iron): ≤ 0.001%, Ti (Titanium): ≤ 0.005%
Odor Weak odor
Packaging Plastic bottle
Product Line EMSURE
Appearance White solid
Density 1.91 g/cm³
Assay Gravimetric
Chemical Name or Material Zirconium(IV) oxide chloride octahydrate
Show More Show Less
Hazard Category H314: Causes severe skin burns and eye damage.