missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) fluoride, low hafnium, 99%, Hf <0.1%, Thermo Scientific Chemicals

$100.10 - $367.18

Chemical Identifiers

CAS 7783-64-4
MDL Number MFCD00011307
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1154214
View Documents
Thermo Scientific Chemicals
01154214
25 g
Each for $100.10
Only null left
 
AA1154222
View Documents
Thermo Scientific Chemicals
01154222
100 g
Each for $367.18
Only null left
 
Description

Description

Zirconium fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-64-4
MFCD00011307

Specifications

600°C (sublimation)
White
25 g
MFCD00011307
14,10175
167.21
Hf <0.1%
Moisture sensitive
Zirconium(IV) fluoride, Low hafnium
7783-64-4
Powder
ZrF4
UN3260
Insoluble in water.
99%
1.59
4.43 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 232-018-1

RTECSNumber : ZH7875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only