CAS RN 7783-64-4
IUPAC Name:
zirconium(4+) tetrafluoride
Synonyms:
zirconium tetrafluoride
Molecular Weight (g/mol):
167.22
Molecular Formula:
F4Zr
InChi Key:
OMQSJNWFFJOIMO-UHFFFAOYSA-J
SMILES:
[F-].[F-].[F-].[F-].[Zr+4]
1
–
4
of
4
results
2
Zirconium fluoride, 99.5%, Thermo Scientific Chemicals
CAS: 7783-64-4 Molecular Formula: F4Zr Molecular Weight (g/mol): 167.21 MDL Number: MFCD00011307
3
Zirconium(IV) fluoride, low hafnium, 99%, Hf <0.1%, Thermo Scientific Chemicals
CAS: 7783-64-4 MDL Number: MFCD00011307
4
Zirconium(IV) fluoride, 98%, Thermo Scientific Chemicals
CAS: 7783-64-4 Molecular Formula: F4Zr MDL Number: MFCD00011307 Synonym: Zirconium tetrafluoride