missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) fluoride, low hafnium, 99%, Hf <0.1%, Thermo Scientific Chemicals

Catalog No. AA1154214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1154214 25 g
AA1154222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1154214 Supplier Thermo Scientific Chemicals Supplier No. 01154214
Only null left

Zirconium fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-64-4
MDL Number MFCD00011307

Specifications

Melting Point 600°C (sublimation)
Color White
Physical Form Powder
Quantity 25 g
Linear Formula ZrF4
UN Number UN3260
Merck Index 14,10175
Solubility Information Insoluble in water.
Formula Weight 167.21
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Hf <0.1%
Refractive Index 1.59
Sensitivity Moisture sensitive
Density 4.43 g/mL
Chemical Name or Material Zirconium(IV) fluoride, Low hafnium
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-018-1
RTECSNumber ZH7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only